Anti-CD44V4 Mouse Monoclonal Antibody [clone: VFF-11]

Supplier: GeneTex
GTX75288
89367-850EA 913.65 CAD
89367-850
Anti-CD44V4 Mouse Monoclonal Antibody [clone: VFF-11]
Antibodies
Mouse monoclonal antibody [VFF-11] to CD44v4

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/50 Immunohistology - Frozen: 1/50 - 1/500 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD44V4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR