Anti-CD44V6 Mouse Monoclonal Antibody [clone: VFF-7]

Supplier: GeneTex

GTX75292
89367-852EA 816.84 CAD
89367-852
Anti-CD44V6 Mouse Monoclonal Antibody [clone: VFF-7]
Antibodies
Mouse monoclonal antibody [VFF-7] to CD44v6

Type: Primary
Antigen: CD44V6
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR