Anti-PTPRC Mouse Monoclonal Antibody [clone: F10-89-4]

Supplier: GeneTex
GTX76583
89366-142EA 822.42 CAD
89366-142
Anti-PTPRC Mouse Monoclonal Antibody [clone: F10-89-4]
Antibodies
Mouse monoclonal antibody [F10-89-4] to CD45

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: F10-89-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR