Anti-PTPRC Mouse Monoclonal Antibody

GTX76088
89365-986EA 397.91 CAD
89365-986
Anti-PTPRC Mouse Monoclonal Antibody
Mouse monoclonal antibody [OX-33] to CD45RA

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR