Anti-CD46 Rabbit Monoclonal Antibody

GTX62538
89364-492EA 556.26 CAD
89364-492
Anti-CD46 Rabbit Monoclonal Antibody
CD46 is a type I membrane protein and a regulatory part of the complement system. This protein has cofactor activity for inactivation of complement components C3b and C4b by serum factor I, which protects the host cell from damage by complement. In addition, CD46 can act as a receptor for the Edmonston strain of measles virus, human herpesvirus-6, and type IV pili of pathogenic Neisseria. Finally, this protein may be involved in the fusion of the spermatozoa with the oocyte during fertilization. CD46 is found in a cluster on chromosome 1q32 with other proteins that are structural components of the complement system.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR