Anti-CD47 Mouse Monoclonal Antibody [clone: MEM-122]

Supplier: GeneTex

GTX29089
89352-562EA 640.8 CAD
89352-562
Anti-CD47 Mouse Monoclonal Antibody [clone: MEM-122]
Antibodies
Mouse monoclonal [MEM - 122] to CD47

FACS: Use at an assay dependant dilution. IHC-Fr: Use at an assay dependant dilution. WB: Use at a dilution of 1/100. Use in non-reducing conditions. This antibody causes platelet aggregation and ATP release from platelet-rich plasma but not from washed platelets, indicating a complement-dependent reaction. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: MEM-122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR