Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]

Supplier: GeneTex
GTX29185
89362-798EA 980.86 CAD
89362-798
Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]
Antibodies
Mouse monoclonal [MEM - 102] to CD48

FACS: Use at an assay dependant concentration. IHC-P: Use at an assay dependant concentration. IP: Use at an assay dependant concentration. WB: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: MEM-102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR