Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]

Supplier: GeneTex
GTX78301
89366-586EA 917.73 CAD
89366-586
Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]
Antibodies
Monoclonal Antibody to CD48 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:1000. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR