Anti-ITGA2 Mouse Monoclonal Antibody [clone: 16B4]

Supplier: GeneTex
GTX75705
89365-880EA 936.47 CAD
89365-880
Anti-ITGA2 Mouse Monoclonal Antibody [clone: 16B4]
Antibodies
Mouse monoclonal antibody [16B4] to Integrin alpha 2

Recommended Starting Dilutions: ELISA: Max Dilution 10 ug/ml Flow Cytometry: Max Dilution 30 ug/ml Immunohistology - Frozen: 30 ug/ml - 40 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone: 16B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR