Anti-ITGA2 Mouse Monoclonal Antibody [clone: AK7]

Supplier: GeneTex
GTX76462
89367-984EA 807.84 CAD
89367-984
Anti-ITGA2 Mouse Monoclonal Antibody [clone: AK7]
Antibodies
Mouse monoclonal antibody [AK7] to Integrin alpha 2

Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR