Anti-CD5 Mouse Monoclonal Antibody [clone: MF7-14.5]

Supplier: GeneTex

GTX74909
89365-626EA 954.51 CAD
89365-626
Anti-CD5 Mouse Monoclonal Antibody [clone: MF7-14.5]
Antibodies
Mouse monoclonal antibody [MF7-14.5] to CD5

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: MF7-14.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR