Anti-CD52 Mouse Monoclonal Antibody [clone: HI186]

Supplier: GeneTex
GTX75945
89353-388EA 730.42 CAD
89353-388
Anti-CD52 Mouse Monoclonal Antibody [clone: HI186]
Antibodies
Mouse monoclonal [HI186] to CD52 (RPE)

FACS: Use 20?l of the suggested working dilution to label 1 x 10^6 cells in 100?l. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR