Anti-ICAM1 Mouse Monoclonal Antibody

Supplier: Genetex
GTX75218
89365-720EA 485.38 CAD
89365-720
Anti-ICAM1 Mouse Monoclonal Antibody
Human CD54 cell surface antigen, also known as ICAM-1, is expressed by many cells following activation by inflammatory mediators.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR