Anti-CD55 Mouse Monoclonal Antibody [clone: BRIC 216]

GTX19772
89360-686EA 569.94 CAD
89360-686
Anti-CD55 Mouse Monoclonal Antibody [clone: BRIC 216]
Avoidance by host tissues of attack by autologous complement proteins is dependent in part on the activities of membrane regulatory factors. One molecule involved in this control is a 70-kD glycoprotein termed decay-accelerating factor (DAF), or CD55. Interruption by CD55 of the complement sequence at an early step in activation effectively halts progression of the cascade and prevents consequent cell injury. In man, CD55 is expressed on the plasma membrane of all cell types that are in intimate contact with plasma complement proteins.This protein recognizes C4b and C3b fragments that condense with cell-surface hydroxyl or amino groups when nascent C4b and C3b are locally generated during C4 and c3 activation. Interaction of daf with cell-associated C4b and C3b polypeptides interferes with their ability to catalyze the conversion of C2 and factor B to enzymatically active C2a and Bb and thereby prevents the formation of C4b2a and C3bBb, the amplification convertases of the complement cascade.CD55 also acts as the receptor for echovirus 7 and related viruses(echoviruses 13, 21, 29 and 33). It is attached to the membrane by a GPI-anchor.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: BRIC 216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR