Anti-NCAM1 Mouse Monoclonal Antibody [clone: ERIC-1]

Supplier: GeneTex
GTX76337
89366-058EA 925.15 CAD
89366-058
Anti-NCAM1 Mouse Monoclonal Antibody [clone: ERIC-1]
Antibodies
Mouse monoclonal antibody [ERIC-1] to NCAM1

Recommended Starting Dilutions: ELISA: Max Dilution 50ng/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR