Anti-CD58 Mouse Monoclonal Antibody [clone: MEM-63]

Supplier: GeneTex

GTX78310
89366-588EA 595.38 CAD
89366-588
Anti-CD58 Mouse Monoclonal Antibody [clone: MEM-63]
Antibodies
Monoclonal Antibody to CD58 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry. Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR