Anti-CGRP Chicken Polyclonal Antibody

Supplier: GeneTex
GTX80742
89366-674EA 887.55 CAD
89366-674
Anti-CGRP Chicken Polyclonal Antibody
Antibodies
Chicken Anti-CGRP Polyclonal Antibody

Immunohistochemistry: 1:100 -1:500 Dilutions listed as a recommendation. Optimal dilution should be determined by investigator. Immunohistochemistry: Antiserum was used on perfusion fixed tissue. Perfusion: 1) calcium-free TyrodeÕs solution, 2) paraformaldehyde-picric acid fixative, and 3) 10% sucrose in PBS as a cryo-protectant. Desired tissues were dissected and stored overnight in 10% sucrose in PBS. Slide-mounted tissue sections were processed for indirect immunofluorescence. Slides were incubated with blocking buffer for 1 hour at room temperature. Primary antiserum was diluted with blocking buffer to the appropriate working concentration. Blocking buffer was removed and slides were incubated for 18-24 hours at 4oC with primary antiserum. Slides were rinsed 3 times and then incubated with secondary antibodies for 1 hour at room temperature. Slides were again rinsed 3 times and coverslipped. Staining was examined using fluorescence microscopy.

Type: Primary
Antigen: CGRP
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR