Anti-MECOM Rabbit Polyclonal Antibody

GTX77822
89366-322EA 684.05 CAD
89366-322
Anti-MECOM Rabbit Polyclonal Antibody
Rabbit Polyclonal Antibody to EVI1

  • WB: Suggested dilution at 0.5-5.0 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: MECOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR