Anti-FGF9 Mouse Monoclonal Antibody [clone: FG9-77]

Supplier: Genetex
GTX10424
89350-266EA 917.95 CAD
89350-266
Anti-FGF9 Mouse Monoclonal Antibody [clone: FG9-77]
Antibodies
Mouse monoclonal [FG9 - 77] to FGF9

Dot: Use at a concentration of 5 ?g/ml. ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 25 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FGF9
Clonality: Monoclonal
Clone: FG9-77
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR