Anti-GHRL Chicken Polyclonal Antibody

GTX15861
89367-444EA 699.81 CAD
89367-444
Anti-GHRL Chicken Polyclonal Antibody
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release from the pituitary. It has an appetite stimulating effect, induces adiposity, and stimulates gastric acid secretion, and it is involved in growth regulation. Ghrelin is synthesized as a preprohormone, then proteolytically processed to yield a 28 amino acid peptide. An interesting and unique modification is imposed on the hormone during synthesis in the form of an n octanoic acid bound to one of its amino acids. This modification is necessary for biologic activity.

Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR