Anti-GHRL Mouse Monoclonal Antibody [clone: Ghr6]

Supplier: GeneTex
GTX13984
89350-892EA 403.99 CAD
89350-892
Anti-GHRL Mouse Monoclonal Antibody [clone: Ghr6]
Antibodies
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release from the pituitary. It has an appetite stimulating effect, induces adiposity, and stimulates gastric acid secretion, and it is involved in growth regulation. Ghrelin is synthesized as a preprohormone, then proteolytically processed to yield a 28 amino acid peptide. An interesting and unique modification is imposed on the hormone during synthesis in the form of an n octanoic acid bound to one of its amino acids. This modification is necessary for biologic activity.

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 13 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GHRL
Clonality: Monoclonal
Clone: Ghr6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR