Anti-CD68 Mouse Monoclonal Antibody

GTX73486
89353-186EA 473.93 CAD
89353-186
Anti-CD68 Mouse Monoclonal Antibody
This antibody reacts with a 110 kD glycoprotein expressed primarily as an intracellular molecule. It stains macrophages in a wide varity of human tissues, including Kuffer's cells and macrophages in the spleen, gut, and lung alveoli and in bone marrow. Peripheral blood monocytes, large lymphocytes and basophils and mast cells are positive but neutrophils stain very weakly.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR