Anti-CD7 Mouse Monoclonal Antibody [clone: CBC.37]

GTX17078
89351-312EA 740.4 CAD
89351-312
Anti-CD7 Mouse Monoclonal Antibody [clone: CBC.37]
CD7 is a 40kD transmembrane, single-chain glycoprotein. CD7 is expressed by majority of peripheral blood T cells, NK cells and all Thymocytes. The presence or absence of CD7 is a useful marker for the classification of T- and NK cell malignancies. The CD7 molecule is absent in a subset of human T cells under certain physiologic conditions. Thus an increase in CD7 negative T cells can be seen in some inflammatory skin disease.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: CBC.37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR