Anti-CD8 Mouse Monoclonal Antibody

Supplier: Genetex
GTX73594
89353-226EA 929.32 CAD
89353-226
Anti-CD8 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR