Anti-CD8 Mouse Monoclonal Antibody [clone: 2ST8.5H7]

Supplier: GeneTex
GTX75282
89365-726EA 1441.26 CAD
89365-726
Anti-CD8 Mouse Monoclonal Antibody [clone: 2ST8.5H7]
Antibodies
Mouse monoclonal antibody [2ST8.5H7] to CD8

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 2ST8.5H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR