Anti-CD8 Mouse Monoclonal Antibody [clone: 341]

Supplier: GeneTex

GTX76645
89353-418EA 692.01 CAD
89353-418
Anti-CD8 Mouse Monoclonal Antibody [clone: 341]
Antibodies
Mouse monoclonal [341] to CD8 Beta (FITC)

For FACS: Use 0 . 5 ug to label 10^6 cells. Protocols are available upon request.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR