Anti-CD8 Rabbit Monoclonal Antibody

GTX62092
89364-312EA 501.38 CAD
89364-312
Anti-CD8 Rabbit Monoclonal Antibody
The T cell coreceptor CD8 is a cell-surface glycoprotein expressed either as a disulfide-linked homodimer of two CD8 alpha monomers, or a heterodimer of CD8 alpha and CD8 beta. These receptors interact with ligands, such as major histocompatibility complex (MHC) class I, on the outside of the cell, with proteins inside the cell, such as the tyrosine kinase p56lck, and possibly with proteins on the same cell-surface . CD8 plays an important role in T cell development in the thymus and T cell activation in the periphery .

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR