Anti-CD8 Rat Monoclonal Antibody [clone: YTS105.18]

Supplier: GeneTex
GTX74642
89365-508EA 1074.35 CAD
89365-508
Anti-CD8 Rat Monoclonal Antibody [clone: YTS105.18]
Antibodies
Rat monoclonal antibody [YTS105.18] to CD8 alpha

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR