Anti-CD80 Mouse Monoclonal Antibody [clone: MEM-233]

Supplier: GeneTex
GTX78330
89353-836EA 710.9 CAD
89353-836
Anti-CD80 Mouse Monoclonal Antibody [clone: MEM-233]
Antibodies
Monoclonal Antibody to CD80 (Human) Allophycocyanin (APC)

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR