Anti-CK8 Rabbit Monoclonal Antibody

GTX61109
89363-826EA 556.26 CAD
89363-826
Anti-CK8 Rabbit Monoclonal Antibody
Keratin polypeptides 8 and 18 (K8/18) are intermediate filament phosphoglycoproteins that are expressed preferentially in glandular epithelia. Cytokeratins play a critical role in differentiation and tissue specialization and function to maintain the overall structural integrity of epithelial cells . Together with actin microfilaments and microtubules, keratin filaments make up the cytoskeletons of vertebrate epithelial cells, forming alpha-helical coiled-coil dimers which associate laterally and end-to-end to form 10-nm diameter filaments .

Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR