Anti-CK8 Mouse Monoclonal Antibody [clone: C-51]

Supplier: GeneTex
GTX22531
89361-274EA 975.6 CAD
89361-274
Anti-CK8 Mouse Monoclonal Antibody [clone: C-51]
Antibodies
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight

For ICC: Use at an assay dependant dilution. For IHC-P: Use at an assay dependant dilution. Perform enzymatic antigen retrieval before commencing with IHC staining protocol. For IP: Use at an assay dependant dilution. For WB: Use at an assay dependant dilution. Predicted molecular weight: 52.5 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: C-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR