Anti-FKBP1A Mouse Monoclonal Antibody [clone: 3B3]

Supplier: GeneTex

GTX84492
89366-794EA 870.24 CAD
89366-794
Anti-FKBP1A Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Mouse monoclonal antibody [3B3] to FKBP1A

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FKBP1A
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR