Germ cell nuclear factor (GCNF), a NR6 Germ Cell Nuclear Factor-Like Receptor, has been shown to affect gene expression in embryogenesis and spermatogenesis and is essential for embryonic survival and normal development. Loss of GCNF causes embryonic lethality and disrupts normal somitogenesis as well as neural-tube and axis formation. GCNF binds as a homodimer to the DR0 DNA response element and has been shown to regulate transcription of genes including protamines1 and 2 and Oct4. GCNF's repression function has been shown to be mediated by interaction with the co-repressors N-COR and SMRT in the absence of ligand. Three protein isoforms, GCNF1, GCNF2a, and GCNF2b, have been documented in humans, and they are generated by differential usage of alternative splice acceptor sites of the fourth and seventh exons. GCNF1, the longest isoform, has a distinct 5' UTR and contains 4 and 26 additional amino acids than the other two forms, respectively. GCNF expression has been documented by in human testis, ovary, prostate, small intesine, and colon. Little or no expression was identified in brain, heart, liver, lung, pancreas, placenta, skeletal muscle, spleen, or thymus.
Type: Primary
Antigen: GCNF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human