Anti-IgE Goat Polyclonal Antibody (Biotin)

Supplier: GeneTex
GTX77206
89353-542EA 431.55 CAD
89353-542
Anti-IgE Goat Polyclonal Antibody (Biotin)
Antibodies
Biotin-conjugated Goat anti-Horse IgE Polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR