Anti-CD9 Rabbit Monoclonal Antibody

GTX62294
89364-384EA 556.26 CAD
89364-384
Anti-CD9 Rabbit Monoclonal Antibody
CD9 belongs to the transmembrane 4 superfamily, also known as tetraspanin (TM4SF) family and is expressed in variety of hematopoietic and epithelial cells . It can form homodimers or homooligomers as well as heterooligomers with other members of the TM4SF family . It can trigger platelet activation and aggregation and modulate cell adhesion and migration. In addition, CD9 appears to promote muscle cell fusion and support. Downregualtion of CD9 has been reported to be associated with tumor progression and metastasis .

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR