Anti-THY1 Rabbit Monoclonal Antibody

GTX62251
89364-368EA 556.26 CAD
89364-368
Anti-THY1 Rabbit Monoclonal Antibody
CD90 (Cluster of Differentiation 90, Thy-1) is a heavily N-glycosylated GPI linked cell surface protein member of the immunoglobulin (Ig) superfamily. As a smallest member of family, CD90 may play a role in cell-to-cell and cell-to-lignand interactions during synaptogensis and other neuron related activities (e.g. neurite outgrowth, nerve regeneration) . In developing nervous tissue, CD90 is involved in axon growth regulation by halting axon growth in mature axon via its interaction with intergin-beta-3 on mature astrocytes. Additionally, CD90 has been linked to apoptosis, adhesion, migration, cancer (including cancer stem cells), and fibrosis . As a major marker for hematopoietic stem cells (HSCs), CD90 mediates differentiation of HSCs. In stem cancer cell research, CD90+ cells in the hepatocellular carcinoma (HCC) have been shown to display tumorgenic capacity .

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR