Anti-THY1 Rabbit Monoclonal Antibody

GTX62252
89364-370EA 556.26 CAD
89364-370
Anti-THY1 Rabbit Monoclonal Antibody
CD90 (Cluster of Differentiation 90, Thy-1) is a heavily N-glycosylated GPI linked cell surface protein member of the immunoglobulin (Ig) superfamily. As a smallest member of family, CD90 may play a role in cell-to-cell and cell-to-ligand interactions during synaptogensis and other neuron related activities (e.g. neurite outgrowth, nerve regeneration) . In developing nervous tissue, CD90 is involved in axon growth regulation by halting axon growth in mature axon via its interaction with intergin-beta-3 on mature astrocytes. Additionally, CD90 has been linked to apoptosis, adhesion, migration, cancer (including cancer stem cells), and fibrosis . As a major marker for hematopoietic stem cells (HSCs), CD90 mediates differentiation of HSCs. In stem cancer cell research, CD90 positive cells in the hepatocellular carcinoma (HCC) have been shown to display tumorigenic capacity .

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR