Anti-CDC14A Mouse Monoclonal Antibody [clone: DCS-291]

GTX10536
89354-220EA 409.37 CAD
89354-220
Anti-CDC14A Mouse Monoclonal Antibody [clone: DCS-291]
Mouse monoclonal [DCS - 291] to Cdc14A

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/2000. Detects a band of approximately 50 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CDC14A
Clonality: Monoclonal
Clone: DCS-291
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR