Anti-CDC25A Mouse Monoclonal Antibody [clone: DCS-121]

Supplier: GeneTex
GTX75690
89367-916EA 1085.88 CAD
89367-916
Anti-CDC25A Mouse Monoclonal Antibody [clone: DCS-121]
Antibodies
Mouse monoclonal antibody [DCS-121] to CDC25A

Recommended Starting Dilutions: Immunoprecipitation: 1 ug/ml - 10 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CDC25A
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR