Anti-CDC42 Rabbit Polyclonal Antibody

GTX100904
89348-744EA 565.14 CAD
89348-744
Anti-CDC42 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to CDC42 (cell division cycle 42 (GTP binding protein, 25kDa))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDC42
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR