Anti-CDC6 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108979
89355-740EA 640.65 CAD
89355-740
Anti-CDC6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cdc6 (cell division cycle 6 homolog (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDC6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR