Anti-CEA Mouse Monoclonal Antibody [clone: 1C10]

GTX10036
89350-592EA 409.01 CAD
89350-592
Anti-CEA Mouse Monoclonal Antibody [clone: 1C10]
Carcino Embryonic Antigen (CEA) is synthesized during development in the fetal gut, and is re expressed in increased amounts in intestinal carcinomas and several other tumors. Antibodies to CEA are useful in identifying the origin of various metastatic adenocarcinomas and in distinguishing pulmonary adenocarcinomas (60 to 70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+).

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: 1C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR