Anti-CEA Mouse Monoclonal Antibody

GTX76561
89353-410EA 521.72 CAD
89353-410
Anti-CEA Mouse Monoclonal Antibody
CEA is a glycoprotein constituent of embryonic endodermal epithelium. It is a complex molecule of 200kD. Specific for CEA, 85 A12 may be used in studying and monitoring primary and secondary tumours of the gastrointestinal tract. Strongly stains tumour cells present in cancerous colon tissue. Only very weak staining of normal tissue observed.

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR