Anti-CENTRIN 1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11257
89357-106EA 922.28 CAD
89357-106
Anti-CENTRIN 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Centrin 1

WB: Use at a dilution of 1/2000, this concentration is recommended using a nuclear extract of the human epitheloid carcinoma HeLa cell line. Predicted molecular weight: 21 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CENTRIN 1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR