Anti-CHK2 Mouse Monoclonal Antibody

GTX70094S
89364-660EA 156.77 CAD
89364-660
Anti-CHK2 Mouse Monoclonal Antibody
Type:
Antigen: CHK2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR