Anti-C-ERBB3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX73146
89353-160EA 599.75 CAD
89353-160
Anti-C-ERBB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to c - erbB - 3 / HER - 3

For IHC-P: Use at an assay dependent dilution. No special pretreatment is required for immuno-histochemical staining of formalin-fixed tissues. For IP: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: C-ERBB3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR