Anti-CG alpha Mouse Monoclonal Antibody [clone: ME.109]

GTX29502
89362-814EA 456.78 CAD
89362-814
Anti-CG alpha Mouse Monoclonal Antibody [clone: ME.109]
Mouse monoclonal [ME.109] to hCG alpha

For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tesed in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CG alpha
Clonality: Monoclonal
Clone: ME.109
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR