Anti-CG Mouse Monoclonal Antibody [clone: G9]

GTX21958
89351-706EA 462.08 CAD
89351-706
Anti-CG Mouse Monoclonal Antibody [clone: G9]
Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.

Type: Primary
Antigen: CG
Clonality: Monoclonal
Clone: G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR