Anti-CHERP Rabbit Polyclonal Antibody

GTX15951
89367-458EA 738.62 CAD
89367-458
Anti-CHERP Rabbit Polyclonal Antibody
The Calcium homoeostasis endoplasmic reticulum protein (CHERP) gene has been recently identified and localized on chromosome 19p13.1. CHERP has high homology to an SR related CTD protein, which is known to interact with the largest subunit of RNA polymerase II. CHERP is an integral endoplasmic reticulum membrane protein is involved in calcium mobilization induced by thrombin. Confocal microscopy revealed that CHERP is associated with Ins(1,4,5)P3 receptor throughout the cytoplasm and perinuclear region in Jurkat T lymphocyte. CHERP Anti sense treatment induced a decrease in CHERP, whichv caused an impaired increase in free cytoplasmic calcium but calcium influx remains unaffected, with some deficient in endoplasmic reticulum calcium stores. In the CHERP depleted Jurkat T lymphocytes the calcium dependent translocation of nuclear factor of activated T cells (NFAT) from cytoplasm to nucleus was also suppressed with significant suppression of cell.

Type: Primary
Antigen: CHERP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR