Anti-Chlamydia Trachomatis Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13943
89359-916EA 849.8 CAD
89359-916
Anti-Chlamydia Trachomatis Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Chlamydia Trachomatis
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR